missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogen Tartrate, Extra Pure, SLR, Fisher Chemical

Product Code. 10666842 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic powder jar
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10666842 1 kg Plastic powder jar 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10666842 Supplier Fisher Chemical Supplier No. P/5480/60
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 868-14-4
Color White
Physical Form Powder/Solid
Quantity 1 kg
Molecular Formula C4H5KO6
MDL Number MFCD00065392,MFCD00071626,MFCD00064206
Synonym potassium bitartrate, l +-potassium hydrogen tartrate, potassium 3-carboxy-2,3-dihydroxypropanoate, monopotassium tartrate, potassium acid tartrate, kaliumtartrat, potassium hydrogentartrate, 1-potassium hydrogentartrate, ksc657e1t, butanedioic acid, 2,3-dihydroxy-2r,3r-, potassium salt 1:1
InChI Key KYKNRZGSIGMXFH-ZVGUSBNCSA-M
SMILES [K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O
IUPAC Name 2,3-dihydroxybutanedioic acid;potassium
Molecular Weight (g/mol) 188.18
PubChem CID 24193652
Formula Weight 188.18g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.