missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogenfluoride, 99%, Honeywell Fluka™

Product Code. p-7120706 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Drum with inliner
Poly Bottle
Unit Size:
2.5kg
25g
500g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15662440

Brand: Honeywell-Fluka 23928325G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

0.99

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-29-9
MDL Number MFCD00011403

Specifications

Boiling Point 180°C
Melting Point 239°C
Quantity 25 g
Linear Formula KHF2
UN Number UN1811
Formula Weight 78.1g/mol
Percent Purity 99%
Packaging Drum with inliner
Density 2.37g/mL at 25°C
Chemical Name or Material Potassium Hydrogenfluoride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.