missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 0.05M in Ethanol, Chem-Lab™

Product Code. 12651201
Change view
Click to view available options
1L; 0.84g/mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12651201 1L; 0.84g/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12651201 Supplier Chem-Lab Supplier No. CL0511071000

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used as a titrant in volumetric solutions. Potassium Hydroxide, 0.05M in Ethanol, Chem-Lab is a laboratory reagent that has 2.806g KOH per liter of ethanol.

Ethanolic potassium hydroxide is an effective conductor of heat and highly flammable. It is used as a dessicant. It is also used in some electrical batteries, such as those used in television remote controls. It is used in the manufacture of soaps, the production of biodiesel and the production of other potassium compounds.

  • Solution may become brown on standing
  • Explosion limit: 3.5–15.0% (V) (ethanol)
  • Ignition temperature: 425°C (ethanol)

Specifications

pH 14 (20°C, H2O)
Color Colorless
Boiling Point 78°C
CAS 1310-58-3, 64-17-5
Melting Point -117°C
Molecular Formula HKO
Concentration 2.806g KOH/l C2H5OH = 0.05 N (±0.0001/20°C)
MDL Number MFCD00003553
Concentration or Composition Notes 2.806g KOH per liter of C2H5OH
Density 0.85g/mL
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Solubility Information Soluble in Water
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Quantity 1 L
SMILES [OH-].[K+]
IUPAC Name potassium;hydroxide
Explosion Limit Lower 3.5% Volume/Upper 15% Volume
Molecular Weight (g/mol) 56.11
Flash Point 12°C (ethanol)
PubChem CID 14797
ChEBI CHEBI:32035
Formula Weight 46.07g/mol
Normality 0.05N
Odor Alcoholic
Vapor Pressure 59hPa at 20°C
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.