missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 90%, Honeywell

Product Code. 15685650 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15685650 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15685650 Supplier Honeywell Research Chemicals Supplier No. 4840161KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1310-58-3
Molecular Formula HKO
Linear Formula KOH
Concentration 90%
MDL Number MFCD00003553
UN Number UN1813
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Quantity 1 kg
SMILES [OH-].[K+]
IUPAC Name potassium;hydroxide
Molecular Weight (g/mol) 56.11
PubChem CID 14797
ChEBI CHEBI:32035
Formula Weight 56.11g/mol
Vapor Pressure 1 mmHg (719°C)
Chemical Name or Material Potassium hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.