missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, ACS Reagent, pellets, ≥85%, Honeywell Fluka™

Product Code. 15662140 Shop All Honeywell Fluka Products
Change view
Klicka för att se tillgängliga alternativ
Quantity:
25 g
500 g
1 kg
2.5 kg
6 x 500 g
12 kg
Förpackningsstorlek:
12kg
1kg
2.5kg
25g
500g
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
15662140 500 g 500g
15632140 25 g 25g
15612140 1 kg 1kg
15622140 2.5 kg 2.5kg
15672140 6 x 500 g Pack of 6
15602140 12 kg 12kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15662140 Leverantör Honeywell-Fluka Leverantörsnummer 221473500G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

ACS Reagent, pellets,≥85%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]

Specifikationer

Linear Formula KOH
UN Number 1813
Quantity 500 g
IUPAC Name potassium hydroxide
Formula Weight 56.11g/mol
Percent Purity ≥85%
Grade ACS Reagent
Vapor Pressure 1mmHg (719°C)
Chemical Name or Material Potassium Hydroxide
compliance-icons compliance-icons
Produktbeteckning
  • Potassium hydroxide
Signalord
  • Danger
Riskkategorier
  • Acute toxicity Category 4
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Faroangivelser
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Skyddsangivelse
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302-if on skin: .
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Kompletterande information
  • MIXTURE LIST-Contain: potassium hydroxide
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.