missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide concentrate, For 1L standard solution, 0.1 M KOH (0.1N), Honeywell Fluka™

Product Code. 15665210 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 Ea.
Unit Size:
Each
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15665210 6 x 1 Ea. Pack of 6
15655210 1 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15665210 Supplier Honeywell Fluka Supplier No. 380706X1EA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

for 1L standard solution, 0.1 M KOH (0.1N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium;hydroxide
SMILES [OH-].[K+]

Specifications

Boiling Point >100°C
CAS Min % ≥80.0000%, ≥10.0000%
CAS Max % <90.0000%, <20.0000%
Linear Formula KOH
UN Number UN1814
Density 1.140 g/cm3
Quantity 6 x 1 Ea.
Formula Weight 56.11
Packaging Ampoule
Physical Form Concentrate
Chemical Name or Material Potassium hydroxide concentrate, For 1L standard solution, 0.1 M KOH (0.1N)
Show More Show Less
compliance-icons
Product Identifier
  • Potassium hydroxide concentrate
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: potassium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.