missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, DILUT-IT Concentrate, 0.5M, J.T. Baker™

Product Code. 10421338 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
For dilution to 1 L
Packaging:
LDPE ampule
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10421338 For dilution to 1 L LDPE ampule Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10421338 Supplier JT Baker Supplier No. 4866

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Prepare fresh, reliable standard solutions quickly and accurately. Potassium Hydroxide, DILUT-IT Concentrate, 0.5M, J.T. Baker™ is a pre-mixed volumetric concentrate for dilution to 1L that is ideal for titrimetry and other forms of wet chemical analysis.

DILUT-IT solutions are relied upon for fast, accurate results in life sciences and pharmaceutical quality control applications. Simply dilute the volumetric concentrate to 1 liter (carefully following instructions provided with the package) and the resulting solution will have an exact normality within ±0.1%.

  • Enhanced productivity: Eliminate expensive, time-consuming steps, such as the selection and analysis of starting reagents, and weighing and standardization of the final solution
  • Space saving: Products require only one-tenth of the space required for ready-made solutions
  • Reduced shipping costs: Spend less on shipping due to the reduced weight and package size of a concentrate

Specifications

Boiling Point 100°C
CAS 1310-58-3
Melting Point 0°C
Molecular Formula HKO
Concentration 0.5mol/L
MDL Number MFCD00003553
Density 1.2g/cm3 at 20°C
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Solubility Information Fully miscible in water
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Quantity For dilution to 1 L
SMILES [OH-].[K+]
IUPAC Name potassium;hydroxide
Molecular Weight (g/mol) 56.11
PubChem CID 14797
ChEBI CHEBI:32035
Formula Weight 56.11g/mol
Grade DILUT-IT
Packaging LDPE ampule
Physical Form Concentrate
Chemical Name or Material Potassium hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.