missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide Pellets, 85%, BAKER ANALYZED, J.T. Baker™

Product Code. 10335870 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
5 kg
Packaging:
HDPE
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10335870 5 kg HDPE 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10335870 Supplier JT Baker Supplier No. 0385.5000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High quality grade pharmaceutical reagent for laboratory and specialized industrial use.

  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability

Specifications

Color White
Boiling Point 1327°C
CAS 1310-58-3
Melting Point 360°C
Molecular Formula HKO
MDL Number MFCD00003553
Density 2.04g/cm3 at 20°C
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Solubility Information 1120g/L H2O
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Quantity 5 kg
SMILES [OH-].[K+]
IUPAC Name potassium hydroxide
Molecular Weight (g/mol) 56.11
PubChem CID 14797
ChEBI CHEBI:32035
Formula Weight 56.11g/mol
Percent Purity ≥85%
Grade BAKER ANALYZED
Odor Odorless
Packaging HDPE
Physical Form Pellet
Chemical Name or Material Potassium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.