missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, puriss. p.a., ≥86% (T), Honeywell Fluka™

Product Code. 15615970 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
25 kg
Unit Size:
1kg
250g
25kg
5kg
This item is not returnable. View return policy

Product Code. 15615970

Brand: Honeywell-Fluka 6037025KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., pellets,≥86% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]

Specifications

Linear Formula KOH
UN Number UN1813
Quantity 25 kg
IUPAC Name potassium hydroxide
Formula Weight 56.11
Percent Purity ≥86% (T)
Vapor Pressure 1 mmHg (719°C)
Chemical Name or Material Potassium hydroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.