missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, Puriss. p.a., Reag. Ph. Eur., ≥85%, pellets, Solstice

Product Code. 15663070 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
6 x 500 g
4 x 5 kg
6 x 1 kg
Unit Size:
1kg
2.5kg
500g
5kg
Pack of 4
Pack of 6
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15663070 6 x 500 g Pack of 6
15633070 500 g 500g
15693060 1 kg 1kg
15603070 2.5 kg 2.5kg
15643070 5 kg 5kg
15653070 6 x 1 kg Pack of 6
15623070 4 x 5 kg Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15663070 Supplier Honeywell-Fluka Supplier No. 306036X500G
  • ONLINE EXCLUSIVE PRICE
    196.00 €
    176.80 € / Pack of 6
    Save 19.20 €
    9% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., Reag. Ph. Eur.,≥85%, pellets

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]

Specifications

Linear Formula KOH
UN Number UN1813
Quantity 6 x 500 g
IUPAC Name potassium;hydroxide
Formula Weight 56.11
Percent Purity ≥85%
Grade puriss. p.a.
Vapor Pressure 1mmHg (719°C)
Chemical Name or Material Potassium Hydroxide, Reag. Ph. Eur.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.