missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide Solution 0.1M (0.1N), (In Methanol), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Description
1L, HDPE plastic bottle
Specifications
Specifications
| pH | 13 |
| Color | Colorless |
| Boiling Point | 64.7°C |
| CAS | 1310-58-3 |
| Molecular Formula | HKO |
| MDL Number | MFCD00003553 |
| Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
| InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
| Quantity | 1 L |
| SMILES | [OH-].[K+] |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction