missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide Solution, volumetric, 0.5M KOH (0.5N), Honeywell Fluka™

Product Code. 15613560 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 mL
2 L
Packaging:
Poly Bottle
Unit Size:
2 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15613560 2 L Poly Bottle 2 L
15623560 500 mL Poly Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15613560 Supplier Honeywell-Fluka Supplier No. 3193682L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Volumetric, 0.5 M KOH (0.5N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium;hydroxide
SMILES [OH-].[K+]

Specifications

Linear Formula KOH
UN Number UN1814
Density 1.025g/mL at 25°C
Quantity 2 L
Formula Weight 56.11g/mol
Packaging Poly Bottle
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide Solution, Volumetric, 0.5M KOH (0.5N)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.