missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, Technical, ≥85%, Honeywell Fluka™

Product Code. 15640810 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly Bottle
Unit Size:
1kg
5kg
Product Code. Quantity Packaging unitSize
15640810 1 kg Poly Bottle 1kg
15650810 5 kg Poly Bottle 5kg
2 options
This item is not returnable. View return policy

Product Code. 15640810

Brand: Honeywell-Fluka 061031KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Technical,≥85%, powder

  • Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
  • Variation in color and physical form is possible
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

pH 14 at 20°C
Color White
Boiling Point 1327°C
Melting Point 360°C
Density 2.04 g/cu.cm
Quantity 1 kg
Percent Purity ≥85%
Grade Technical
Packaging Poly Bottle
Vapor Pressure 1mmHg (719°C)
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.