missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Potassium indigotrisulfonate

Product Code. 10206990 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10206990 5 g Glass bottle 5g
10533961 1 g Glass bottle 1g
11976311 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10206990 Supplier Thermo Scientific Chemicals Supplier No. 215170050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 67627-18-3 | C16H7K3N2O11S3 | 616.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67627-18-3
Molecular Formula C16H7K3N2O11S3
Molecular Weight (g/mol) 616.71
MDL Number MFCD00013160
InChI Key XOSMXDUITYWYGR-JRYLAINFSA-K
Synonym potassium indigotrisulfonate, indigotrisulfonic acid tripotassium salt, indigotrisulfonate potassium salt, indigotrisulfonic acid potassium salt, unii-5zza8n0abt, 5zza8n0abt, tripotassium indigotrisulfonate, tri-potassium indigotrisulfonate, tripotassium indigo-5,5',7-trisulfonate, tri-potassium indigo-5,5',7-trisulfonate
PubChem CID 6364606
IUPAC Name tripotassium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5,7-disulfonate
SMILES [K+].[K+].[K+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1/NC2=C(C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O)C1=O

Specifications

Packaging Glass bottle
Quantity 5 g
Formula Weight 616.71
Chemical Name or Material Potassium indigotrisulfonate
compliance-icons
Product Identifier
  • Potassium indigotrisulfonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.