missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodate, 98%, extra pure

Product Code. 10571831 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10571831 100 g Glass bottle 100g
10388140 500 g Glass bottle 500g
10083903 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10571831 Supplier Thermo Scientific Acros Supplier No. 196741000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7758-05-6 | IKO3 | 214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-05-6 , 7732-18-5
Molecular Formula IKO3
Molecular Weight (g/mol) 214
MDL Number MFCD00011406
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
IUPAC Name potassium iodate
SMILES [K+].[O-][I](=O)=O

Specifications

Melting Point 560°C
Color White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 97.5% min. (Iodometry)
Linear Formula KIO3
UN Number 1479
Merck Index 15, 7763
Solubility Information Solubility in water: soluble
Formula Weight 214
Percent Purity 98%
Grade Extra Pure
Loss on Drying 0.5% max.
Packaging Glass bottle
Specific Gravity 3.93
Density 3.9300g/mL
Chemical Name or Material Potassium iodate, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium iodate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.