missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate, ACS Reagent, 99.5%, Honeywell™ Fluka™

Product Code. 15662010 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
12 kg
4 x 2.5 kg
Packaging:
Drum with Inliner
Poly Bottle
Unit Size:
100g
12kg
2.5kg
500g
5g
Pack of 4
Product Code. Quantity Packaging unitSize
15662010 500 g Poly Bottle 500g
15672010 5 g Poly Bottle 5g
15622010 100 g Poly Bottle 100g
15642010 2.5 kg Poly Bottle 2.5kg
15652010 4 x 2.5 kg Poly Bottle Pack of 4
15632010 12 kg Drum with Inliner 12kg
6 options
This item is not returnable. View return policy

Product Code. 15662010

Brand: Honeywell-Fluka 215929500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACS Reagent, 99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5-6-7758
Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
MDL Number MFCD00011406
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710
IUPAC Name potassium;iodate
SMILES [O-]I(=O)=O.[K+]

Specifications

Melting Point 560°C
Color Colorless
pH 5 to 8 (at 20°C)
Physical Form Solid
Quantity 500 g
Linear Formula KIO3
UN Number UN1479
Formula Weight 214g/mol
Percent Purity 99.50%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Crystals
Density 3.93g/mL at 25°C; Bulk Density: ca. 2400kg/cu.m
Chemical Name or Material Potassium Iodate
Show More Show Less
compliance-icons
Product Identifier
  • Potassium iodate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : Potassium iodate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.