missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate, Honeywell Fluka™

Product Code. 15613000 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
25 kg
Unit Size:
100g
1kg
250g
25kg
Product Code. Quantity unitSize
15613000 25 kg 25kg
15682990 100 g 100g
15603000 250 g 250g
15692990 1 kg 1kg
4 options
This item is not returnable. View return policy

Product Code. 15613000

Brand: Honeywell-Fluka 3031425KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., 99.7-100.4%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5-6-7758
Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
MDL Number MFCD00011406
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710
IUPAC Name potassium;iodate
SMILES [O-]I(=O)=O.[K+]

Specifications

Quantity 25 kg
Linear Formula KIO3
UN Number 1479
Formula Weight 214g/mol
Percent Purity 99.7 to 104%
Grade puriss. p.a.
Density 3.93g/mL at 25°C
Chemical Name or Material Potassium Iodate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.