missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodate, 99.4+%, ACS reagent

Product Code. 10697201 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10697201 500 g Plastic bottle 500g
10762212 100 g Plastic bottle 100g
10732204 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10697201 Supplier Thermo Scientific Acros Supplier No. 418245000
  • ONLINE EXCLUSIVE PRICE
    191.00 €
    185.00 € / 500g
    Save 6.00 €
    3% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7758-05-6 | IKO3 | 214.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5-6-7758
Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
MDL Number MFCD00011406
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710
IUPAC Name potassium;iodate
SMILES [O-]I(=O)=O.[K+]

Specifications

Color White
pH 5.0 to 8.0 (5% soln. at 25°C)
Physical Form Crystalline Powder and/or Crystals
Quantity 500 g
Assay Percent Range 99.4+%
Linear Formula KIO3
Merck Index 15, 7763
Formula Weight 214
Percent Purity 99.4 to 100.4% (Iodometry)
Grade ACS Reagent
Packaging Plastic bottle
Decomposition Information 560°C
Density 3.9300g/mL
Chemical Name or Material Potassium iodate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.