missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodate solution, 0.017M, Solstice

Product Code. 15674140
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15674140 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15674140 Supplier Honeywell-Fluka Supplier No. 342731L
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
Estimated Shipment: 01-02-2027
Add to basket
Edge
This item is not returnable. View return policy

Volumetric, 1/60 M KIO3 (0.1N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Specifications

CAS 5-6-7758
Quantity 1 L
Molecular Formula IKO3
Linear Formula KIO3
MDL Number MFCD00011406
UN Number NONH for all modes of transport
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
SMILES [O-]I(=O)=O.[K+]
IUPAC Name potassium;iodate
Molecular Weight (g/mol) 214.00
PubChem CID 23665710
Formula Weight 214g/mol
Concentration or Composition (by Analyte or Components) 0.017M
Chemical Name or Material Potassium iodate solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.