missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodide, 99.995%, (trace metal basis), extra pure

Product Code. 10562981 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10562981 50 g Glass bottle 50g
10557183 10 g Glass bottle 10g
10338753 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10562981 Supplier Thermo Scientific Acros Supplier No. 193790500
  • ONLINE EXCLUSIVE PRICE
    266.00 €
    249.00 € / 50g
    Save 17.00 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7681-11-0 | IK | 166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1330.0°C
Melting Point 680.0°C
Color White
Physical Form Crystals or Powder
Quantity 50 g
Packaging Glass bottle
Assay Percent Range 99.995%
Linear Formula KI
Fieser 06,227; 11,440
Merck Index 15, 7764
Solubility Information Solubility in water: 1.43 kg/l. Other solubilities: 45g/L ethanol
Formula Weight 166
Percent Purity 99.995%
Grade Trace Metal Basis
Chemical Name or Material Potassium iodide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.