missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, Certified AR for Analysis, Fisher Chemical™

Product Code. 10264060 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
3 kg
Unit Size:
100g
250g
3kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10264060 3 kg 3kg
10607232 100 g 100g
10793801 250 g 250g
10703811 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10264060 Supplier Fisher Chemical Supplier No. P/5880/63
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

IK, CAS Number-7681-11-0, kali iodide, asmofug e, thyro-block, pima, potassium monoiodide, knollide, kisol, potassium diiodide, potassium iodide ki, potassium iodide, 500g, 1330 deg.C, CHEBI:8346, White, 166g/mol, NLKNQRATVPKPDG-UHFFFAOYSA-M, potassium;iodide, 680 deg.C, 11405, 166.003

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number 11405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1330°C
Melting Point 680°C
Color White
pH 6 to 8
Physical Form Solid
Quantity 3 kg
Formula Weight 166g/mol
Packaging Plastic powder jar
Vapor Pressure 1mmHg 745
compliance-icons
Product Identifier
  • Potassium iodide
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.