missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, Extra Pure, SLR, Fisher Chemical™

Product Code. 10489180 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
250g
500g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10489180 500 g 500g
11468772 100 g 100g
10766491 250 g 250g
10132060 1 kg 1kg
10409570 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10489180 Supplier Fisher Chemical Supplier No. P/5840/53
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7681-11-0 | Molecular Formula: IK | Molecular Weight: 166.003 g/mol

Potassium Iodide, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number 11405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1330°C
Melting Point 680°C
Color White
pH 6 to 8
Physical Form Solid
Quantity 500 g
Formula Weight 166g/mol
Packaging Plastic powder jar
Vapor Pressure 1mmHg 745
compliance-icons
Product Identifier
  • Potassium iodide
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.