missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodide, Puriss., meets analytical spec. of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance), Honeywell Fluka™
BIOPHARMA

Product Code. 15610670 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
6 x 250 g
25 kg
50 kg
6 x 1 kg
Unit Size:
1kg
250g
25kg
50kg
5kg
Pack of 6
Product Code. Quantity unitSize
15610670 250 g 250g
15600670 1 kg 1kg
15660670 6 x 250 g Pack of 6
15640670 5 kg 5kg
15650670 6 x 1 kg Pack of 6
15620670 25 kg 25kg
15630670 50 kg 50kg
7 options
This item is not returnable. View return policy

Product Code. 15610670

Brand: Honeywell-Fluka 03124250G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Quantity 250 g
Linear Formula KI
UN Number NONH for all modes of transport
Formula Weight 166g/mol
Percent Purity 99 to 100.5% (calc. to the dried substance)
Grade puriss.
Packaging Plastic Bottle
Vapor Pressure 1mmHg (745°C)
Chemical Name or Material Potassium Iodide, Meets analytical specification of Ph.Eur. BP, USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.