missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodide, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥99.5%, Solstice

Product Code. 15643000 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
5 kg
6 x 100 g
6 x 250 g
6 x 500 g
4 x 5 kg
50 kg
6 x 1 kg
Unit Size:
100g
1kg
250g
500g
50kg
5kg
Pack of 4
Pack of 6
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15643000 250 g 250g
15623000 100 g 100g
15663000 500 g 500g
15693000 6 x 100 g Pack of 6
15633000 1 kg 1kg
15613010 6 x 250 g Pack of 6
15623010 6 x 500 g Pack of 6
15683000 5 kg 5kg
15603010 6 x 1 kg Pack of 6
15653000 4 x 5 kg Pack of 4
15673000 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 15643000 Supplier Honeywell-Fluka Supplier No. 30315250G
  • ONLINE EXCLUSIVE PRICE
    185.00 €
    55.95 € / 250g
    Save 129.05 €
    69% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Puriss. p.a., Reag. ISO, Reag. Ph. Eur.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Quantity 250 g
Linear Formula KI
UN Number NONH for all modes of transport
Formula Weight 166g/mol
Percent Purity ≥99.5%
Grade ACS/puriss. p.a.
Vapor Pressure 1mmHg (745°C)
Chemical Name or Material Potassium Iodide, Reag. Ph. Eur. reag. ISO
Product Identifier
  • Potassium iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.