missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, TraceSELECT™, ≥99.999% (metals basis), Honeywell™ Fluka™

Product Code. 15663060 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
Packaging:
Poly bottle
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 15663060

Brand: Honeywell-Fluka 30533100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 1323°C (1013hPa)
Melting Point 681°C
CAS 7681-11-0
Color Colorless
pH 6.0 to 9.0 (at 20°C)
Physical Form Crystalline
Quantity 100 g
Molecular Formula IK
Linear Formula KI
MDL Number MFCD00011405
UN Number NONH for all modes of transport
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES [K+].[I-]
IUPAC Name potassium;iodide
Molecular Weight (g/mol) 166.003
PubChem CID 4875
ChEBI CHEBI:8346
Formula Weight 166g/mol
Percent Purity ≥99.999% (metals basis)
Grade TraceSELECT
Odor Odorless
Packaging Poly bottle
Vapor Pressure 1mmHg (745°C)
Density ∽3.12g/cu.cm at 20°C
Chemical Name or Material Potassium Iodide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.