missing translation for 'onlineSavingsMsg'
Learn More

Potassium metabisulfite, 95+%

Product Code. 11334958
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334958 2 kg 2kg
11324958 100 g 100g
11314958 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334958 Supplier Thermo Scientific Alfa Aesar Supplier No. 043908.A3
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16731-55-8 | K2O5S2 | 222.31 g/mol

It serves as a component of certain photographic developers and its solution is used in photographic fixing, in the textile industry for dyeing and cotton printing. It is utilized to precipitate gold from solution in aqua regia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16731-55-8
Molecular Formula K2O5S2
Molecular Weight (g/mol) 222.31
MDL Number MFCD00167605
InChI Key RWPGFSMJFRPDDP-UHFFFAOYSA-L
Synonym potassium metabisulfite, potassium disulfite, potassium pyrosulfite, dipotassium disulfite, dipotassium pyrosulfite, dipotassium metabisulfite, dipotassium disulphite, unii-65oe787q7w, ccris 1427, potassium disulfite k2s2o5
PubChem CID 28019
IUPAC Name dipotassium sulfinatosulfonate
SMILES [K+].[K+].[O-]S(=O)S([O-])(=O)=O

Specifications

Melting Point 190°C (decomposition)
Color White
Physical Form Powder
Quantity 2 kg
Assay Percent Range >99%
Solubility Information Soluble in water. Insoluble in ethanol.
Formula Weight 222.32
Odor Pungent
Sensitivity Air Sensitive
Chemical Name or Material Potassium metabisulfite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.