missing translation for 'onlineSavingsMsg'
Learn More

Potassium Metabisulfite NF/FCC Granular, Macron Fine Chemicals™

Product Code. 15558384
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly
Unit Size:
2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15558384 2.5 kg Poly 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15558384 Supplier Avantor Macron Supplier No. 700006

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 16731-55-8
Physical Form Granules
Quantity 2.5 kg
Molecular Formula K2O5S2
MDL Number MFCD00167605
Synonym potassium metabisulfite, potassium disulfite, potassium pyrosulfite, dipotassium disulfite, dipotassium pyrosulfite, dipotassium metabisulfite, dipotassium disulphite, unii-65oe787q7w, ccris 1427, potassium disulfite k2s2o5
InChI Key RWPGFSMJFRPDDP-UHFFFAOYSA-L
SMILES [K+].[K+].[O-]S(=O)S([O-])(=O)=O
IUPAC Name dipotassium sulfinatosulfonate
Molecular Weight (g/mol) 222.31
PubChem CID 28019
Formula Weight 222.33
Grade FCC/NF
Packaging Poly
Chemical Name or Material Potassium Metabisulfite
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • di-Potassium disulphite
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : di-Potassium disulphite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.