missing translation for 'onlineSavingsMsg'
Learn More

Potassium methoxide, 90+%

Product Code. 11347847
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347847 50 g 50g
11337847 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347847 Supplier Thermo Scientific Alfa Aesar Supplier No. 014261.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 865-33-8 | CH3KO | 70.132 g/mol

Potassium methoxide is used as a chemical intermediate. It is also used in biodiesel production and in fuel additives. It acts as a catalyst for the preparation of methyl formate and an alkaline catalyst for dimethyl formamide. Further, it is used as versatile reagents in chemical reactions such as condensation, esterification, alkoxylation and etherification, Claisen condensation, Wolf-Kishner reduction and Stobbe reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-33-8
Molecular Formula CH3KO
Molecular Weight (g/mol) 70.132
MDL Number MFCD00012416
InChI Key BDAWXSQJJCIFIK-UHFFFAOYSA-N
Synonym potassium methoxide, potassium methylate, potassium methanolate, methanol, potassium salt, unii-45myq0gwgb, 45myq0gwgb, potassiummethoxide, methanol, potassium salt 1:1, kome, meok
PubChem CID 23664618
IUPAC Name potassium;methanolate
SMILES C[O-].[K+]

Specifications

Physical Form Powder
Quantity 50 g
Assay Percent Range >90%
Solubility Information Soluble in methanol and ethanol.
Formula Weight 70.14
Sensitivity Air and moisture sensitive
Chemical Name or Material Potassium methoxide
compliance-icons compliance-icons
Product Identifier
  • Potassium methoxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Self-heating substance Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H251-Self-heating: may catch fire.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P235-Keep cool.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P420-Store away from other materials.
Supplemental information
  • MIXTURE LIST-Contain: Methanol, potassiumsalt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.