missing translation for 'onlineSavingsMsg'
Learn More

Potassium methoxide, pure, 0.1N solution in toluene/methanol, AcroSeal™

Product Code. 10443160 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10443160 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10443160 Supplier Thermo Scientific Acros Supplier No. 429101000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 865-33-8 | CH3KO | 70.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 865-33-8, 67-56-1, 108-88-3
Quantity 100 mL
Molecular Formula CH3KO
Linear Formula CH3OK
MDL Number MFCD00012416
Flash Point 7°C
Synonym potassium methoxide, potassium methylate, potassium methanolate, methanol, potassium salt, unii-45myq0gwgb, 45myq0gwgb, potassiummethoxide, methanol, potassium salt 1:1, kome, meok
Solubility Information Solubility in water: may decompose
InChI Key BDAWXSQJJCIFIK-UHFFFAOYSA-N
SMILES C[O-].[K+]
IUPAC Name potassium;methanolate
Molecular Weight (g/mol) 70.13
PubChem CID 23664618
Formula Weight 70.13
Grade Pure
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.85
Density 0.8500g/mL
Chemical Name or Material Potassium methoxide, Pure, 0.1N solution in toluene/methanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium methoxide, 0.1N solution in toluene / methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.