missing translation for 'onlineSavingsMsg'
Learn More

Potassium Nitrate Solution, 10%, Reagecon™
BIOPHARMA

Product Code. 11758455 Shop All Reagecon Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 11758455

Brand: Reagecon EFSKNO

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use in the outer chamber of double junction pH electrodes when using media that contains copper, lead, silver or mercury ions. Potassium Nitrate Solution, 10%, Reagecon™ is a standard solution of KNO3. It is also used as a standard in the testing of dairy products.

The glass layer and reference junctions of pH electrodes need to be kept moist when not in use to remain operable and functioning at optimal levels. Potassium Chloride cannot be used with media that contains Hg++, Cu++, Ag+ or Pb++ ions, as they form insoluble precipitates with Cl that will block the diaphragm. Potassium nitrate is a double junction bridge solution that can be used in this case.

Specifications

Chemical Name or Material Potassium Nitrate
CAS 7757-79-1
Color Colorless
Concentration 10% w/v
For Use With (Equipment) Double junction pH electrodes
Physical Form Liquid
Quantity 100 mL
Molecular Formula KNO3
MDL Number MFCD00011409
Solubility Information Miscible in Water
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
SMILES [K+].[O-][N+]([O-])=O
IUPAC Name potassium nitrate
Molecular Weight (g/mol) 101.102
Show More Show Less
compliance-icons
Product Identifier
  • ELECTRODE FILLING SOLUTION 10% POTASSIUM NITRATE
Signal Word
  • Danger
Hazard Category
  • Oxidising liquid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use to extinguish.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : potassium nitrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.