missing translation for 'onlineSavingsMsg'
Learn More

Potassium osmate(VI) dihydrate, 51.0-52.0% Os

Product Code. 10194210 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
500mg
This item is not returnable. View return policy

Product Code. 10194210

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10022-66-9 | K2O4Os·2H2O | 368.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10022-66-9
Molecular Formula K2O4Os·2H2O
Molecular Weight (g/mol) 368.42
MDL Number MFCD00149919
InChI Key DGODWNOPHMXOTR-UHFFFAOYSA-N
Synonym potassium osmate vi dihydrate, potassium dioxidodioxoosmium dihydrate, potassium osmate dihydrate, unii-5m2bwq2tyj, 5m2bwq2tyj, osmate oso42-, dipotassium, dihydrate, t-4, dipotassium tetrahydroxodioxoosmate, potassium osmate k2oso2 oh 4, potassium osmate k2oso4 , dihydrate
PubChem CID 53393272
IUPAC Name dipotassium;dioxido(dioxo)osmium;dihydrate
SMILES O.O.[O-][Os](=O)(=O)[O-].[K+].[K+]

Specifications

Color Violet
Physical Form Crystalline Powder
Quantity 500 mg
Linear Formula K2OsO4·2H2O
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol and ether
Formula Weight 368.42
Percent Purity 51.0 to 52% Os
Packaging Glass bottle
Chemical Name or Material Potassium osmate(VI) dihydrate, 51.0-52.0% Os

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.