missing translation for 'onlineSavingsMsg'
Learn More

Potassium oxalate monohydrate, Puriss., ≥99%, Honeywell

Product Code. 15692740 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
50 kg
Packaging:
Fiberboard Box
Unit Size:
50kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15692740 50 kg Fiberboard Box 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15692740 Supplier Honeywell Research Chemicals Supplier No. 2541350KG
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 6487-48-5
Quantity 50 kg
Molecular Formula C2H2K2O5
MDL Number MFCD00150033
Beilstein 3752576
Synonym potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
InChI Key QCPTVXCMROGZOL-UHFFFAOYSA-L
SMILES O.[K+].[K+].[O-]C(=O)C([O-])=O
IUPAC Name dipotassium hydrate oxalate
Molecular Weight (g/mol) 184.23
PubChem CID 2724193
Percent Purity >99%
Grade puriss.
Packaging Fiberboard Box
Chemical Name or Material Potassium oxalate monohydrate, Puriss., ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.