missing translation for 'onlineSavingsMsg'
Learn More

Potassium Perchlorate, 99+%, Extra Pure, SLR, Fisher Chemical

Product Code. 10244300 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10244300 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 10244300 Supplier Fisher Chemical Supplier No. P/6400/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Specifications

Melting Point 400°C
CAS 7778-74-7
Color White
pH 5.0 to 7.0
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula ClKO4
MDL Number MFCD00011362
Synonym potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp
InChI Key YLMGFJXSLBMXHK-UHFFFAOYSA-M
SMILES [K+].[O-][Cl](=O)(=O)=O
IUPAC Name potassium;perchlorate
Molecular Weight (g/mol) 138.54
PubChem CID 516900
Formula Weight 138.55g/mol
Packaging Plastic powder jar
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.