missing translation for 'onlineSavingsMsg'
Learn More

Potassium periodate, 99.5%, extra pure

Product Code. 10296870 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10296870

Brand: Thermo Scientific Acros 198395000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-21-8 | IKO4 | 230 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-21-8
Molecular Formula IKO4
Molecular Weight (g/mol) 230
InChI Key FJVZDOGVDJCCCR-UHFFFAOYSA-M
Synonym potassium periodate, potassium metaperiodate, unii-q3loc6p66l, q3loc6p66l, periodic acid hio4 , potassium salt, potassium ion io4, ssmicroaea 1/4 o, acmc-20ajue, ssmicroaea 1/4 oni, periodic acid hio4 , potassium salt 1:1
PubChem CID 516896
IUPAC Name potassium;periodate
SMILES [O-]I(=O)(=O)=O.[K+]

Specifications

Melting Point 582.0°C
Color White
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99.4% min. (Iodometry)
Linear Formula KIO4
Fieser 01,809; 02,311; 04,373; 05,507; 06,455
Merck Index 15,7775
Solubility Information Solubility in water: 0.42 g/100mL (20°C).
Formula Weight 230
Percent Purity 99.5%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Potassium periodate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium periodate
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 1
  • Skin corrosion/irritation Category 1 C
  • Specific target organ toxicity Category 1
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use to extinguish.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.