Learn More
Potassium Permanganate Solution, Ph. Eur., Reagecon™
Choose from a wide range of reagent and standard solutions. Reagecon™ Potassium Permanganate Solution, Ph. Eur. is manufactured and tested in compliance with the Eur. Ph.
Brand: Reagecon 1070902
Code : A0
Additional Details : Weight : 1.05100kg Transport : UN number : 3082 Chem class : 9 Pack group : III
Description
- Reagents and Standard solutions in Chapters 2 and 4 of the current Ph. Eur.
- All are manufactured and tested in compliance with the Ph. Eur.
- Supplied with a Certificates of Analysis
- Lot No. and expiry date are stated on all product labels
Specifications
1 L | |
Ph. Eur. |
Safety and Handling
- POTASSIUM PERMANGANATE SOLUTION
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Reproductive toxicity Category 2
Hazard Statement
- H361d-Suspected of damaging the unborn child.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P391-Collect spillage.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : potassium permanganate