missing translation for 'onlineSavingsMsg'
Learn More

Potassium Pyroantimonate Solution, Ph. Eur. Grade, Reagecon™
BIOPHARMA

Product Code. 11767966 Shop All Reagecon Products
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
This item is not returnable. View return policy

Product Code. 11767966

Brand: Reagecon 1071301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Choose from a wide range of reagent and standard solutions. Reagecon™Potassium Pyroantimonate Solution, Ph. Eur. Grade is manufactured and tested in compliance with the Ph. Eur.

European Pharmacopoeia Solutions

  • Reagents and Standard solutions in Chapters 2 and 4 of the current Ph. Eur.
  • All are manufactured and tested in compliance with the Ph. Eur.
  • Supplied with a Certificates of Analysis
  • Lot No. and expiry date are stated on all product labels

Specifications

Quantity 125 mL
compliance-icons compliance-icons
Product Identifier
  • POTASSIUM PYROANTIMONATE SOLUTION
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : potassium hydroxide,potassium hexahydroxoantimonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.