missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate, dibasic, 98+%, ACS reagent

Product Code. 13438578 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic Drum
Plastic bottle
Unit Size:
10kg
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13438578 10 kg Plastic Drum 10kg
10546982 5 g Glass Bottle 5g
10331211 500 g Plastic bottle 500g
10184333 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 13438578 Fornitore Thermo Scientific Acros N. del fornitore 424190100
  • ONLINE EXCLUSIVE PRICE
    790.00 €
    737.00 € / 10kg
    Save 53.00 €
    6% Off
 Restricted Item
In Stock
Aggiungi al carrello
Edge
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 7758-11-4 | HK2O4P | 174.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.18
MDL Number MFCD00011383
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifica

Melting Point 340.0°C
Color White
pH 8.5 to 9.6 (5% soln. at 25°C)
Physical Form Fine Crystalline Powder
Quantity 10 kg
Loss on Drying 1.0% max. (105°C)
Packaging Plastic Drum
Assay Percent Range 98+%
Linear Formula K2HPO4
Merck Index 15,7779
Solubility Information Solubility in water: 1600g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 174.18
Percent Purity ≥98%
Grade ACS Reagent
Chemical Name or Material Potassium phosphate, dibasic
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.