missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Dibasic, 99%, (Crystalline), MP Biomedicals

Product Code. 11447202 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 kg
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447202 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11447202 Supplier MP Biomedicals Supplier No. 0215194505
 Restricted Item
Estimated Shipment: 18-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Anhydrous potassium phosphate dibasic suitable for various laboratory applications.

Specifications

CAS 11-4-7758
Physical Form Crystalline
Quantity 5 kg
Assay Percent Range 99%
Molecular Formula HK2O4P
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
SMILES OP(=O)([O-])[O-].[K+].[K+]
IUPAC Name dipotassium;hydrogen phosphate
Molecular Weight (g/mol) 174.17
PubChem CID 24450
ChEBI CHEBI:32031
Formula Weight 174.2
Percent Purity 100%
Chemical Name or Material Potassium Phosphate Dibasic, Anhydrous
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.