missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Dibasic,≥99% (T), puriss. p.a., ACS reagent, Solstice

Product Code. 15665960
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15665960 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15665960 Supplier Honeywell-Fluka Supplier No. 603561KG
  • ONLINE EXCLUSIVE PRICE
    210.00 €
    85.03 € / 1kg
    Save 124.97 €
    59% Off
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, anhydrous,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 11-4-7758
Physical Form Powder
Quantity 1 kg
Molecular Formula HK2O4P
Linear Formula K2HPO4
MDL Number MFCD00011383
UN Number NONH for all modes of transport
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
SMILES OP(=O)([O-])[O-].[K+].[K+]
IUPAC Name dipotassium;hydrogen phosphate
Molecular Weight (g/mol) 174.17
PubChem CID 24450
ChEBI CHEBI:32031
Formula Weight 174.18g/mol
Percent Purity ≥99% (T)
Grade ACS
Chemical Name or Material Potassium phosphate dibasic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.