missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate, Dibasic, Endotoxin Tested, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 15548104 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
Unit Size:
2.5kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15548104 2.5 kg Poly Bottle 2.5kg
15538104 500 g Poly Bottle 500g
15558104 5 kg Poly Pail 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15548104 Supplier Avantor J.T.Baker Supplier No. 325005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Quantity 2.5 kg
Formula Weight 174.18
Grade Multi-Compendial/USP
Packaging Poly Bottle
Chemical Name or Material Potassium Phosphate, Dibasic, Endotoxin Tested
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.