missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Dibasic Trihydrate, MP Biomedicals™

Product Code. 11420572 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420572 500 g 500g
11410572 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420572 Supplier MP Biomedicals Supplier No. 02191432.5
 Restricted Item
Estimated Shipment: 18-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Widely used in molecular biology, biochemistry and chromatography. Potassium Phosphate Dibasic Trihydrate, MP Biomedicals is a reagent with high buffering capacity. It is typically used as a component in microorganism culture media and in phosphate buffer

Potassium phosphate occurs in several forms: monobasic, dibasic and tribasic. Most pH neutral potassium phosphate buffer solutions consist of mixtures of the monobasic and dibasic forms to varying degrees, depending on the desired pH.

  • It is typically used as a component for a wide variety of media used in the culture of microorganisms
  • It is a component in phosphate buffered saline (PBS)
  • In addition to helping maintain pH, it supplies essential phosphate

Chemical Identifiers

CAS 16788-57-1
Molecular Formula H7K2O7P
Molecular Weight (g/mol) 228.22
MDL Number MFCD00149926
InChI Key XQGPKZUNMMFTAL-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate
PubChem CID 16217523
IUPAC Name dipotassium trihydrate hydrogen phosphate
SMILES O.O.O.[K+].[K+].OP([O-])([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 500 g
Solubility Information Soluble in water (100mg/mL); Insoluble to slightly soluble in ethanol; insoluble in alcohol.
Formula Weight 228.18g/mol
Percent Purity 99.8%
Grade Not Available
Chemical Name or Material Potassium Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.