missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Dibasic Trihydrate, Puriss. p.a., ≥99.0% (T), Honeywell™ Fluka™

Product Code. 15635960 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Poly bottle
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15635960 250 g Poly bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15635960 Supplier Honeywell-Fluka Supplier No. 60349250G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a.,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 16788-57-1
Color White
Physical Form Solid
Quantity 250 g
Molecular Formula H7K2O7P
Linear Formula K2HPO4 · 3H2O
MDL Number MFCD00149926
UN Number NONH for all modes of transport
Synonym dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate
InChI Key XQGPKZUNMMFTAL-UHFFFAOYSA-L
SMILES O.O.O.[K+].[K+].OP([O-])([O-])=O
IUPAC Name dipotassium trihydrate hydrogen phosphate
Molecular Weight (g/mol) 228.22
PubChem CID 16217523
Formula Weight 228.22g/mol
Percent Purity ≥99.0% (T)
Packaging Poly bottle
Chemical Name or Material Potassium phosphate dibasic trihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.