missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate, Dibasic, Ultrapure Bioreagent, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly bottle
Unit Size:
Each
Description
- For Liquid Chromatography and Molecular Biology applications
Specifications
Specifications
| CAS | 11-4-7758 |
| Physical Form | Powder |
| Quantity | 2.5 kg |
| Molecular Formula | HK2O4P |
| Synonym | dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate |
| InChI Key | ZPWVASYFFYYZEW-UHFFFAOYSA-L |
| SMILES | OP(=O)([O-])[O-].[K+].[K+] |
| IUPAC Name | dipotassium;hydrogen phosphate |
| Molecular Weight (g/mol) | 174.17 |
| PubChem CID | 24450 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction