missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate, monobasic, 99%, for analysis

Product Code. 12685087 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12685087 1 kg 1kg
12665087 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12685087 Supplier Thermo Scientific Acros Supplier No. 447670010
  • ONLINE EXCLUSIVE PRICE
    101.00 €
    94.20 € / 1kg
    Save 6.80 €
    6% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7778-77-0 | H2KO4P | 136.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium;dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Melting Point 253.0°C
Quantity 1 kg
Packaging Plastic bottle
Assay Percent Range 99%
Linear Formula KH2PO4
Merck Index 15,778
Solubility Information Solubility in water: 222g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 136.09
Percent Purity 99%
Grade Analytical
Chemical Name or Material Potassium phosphate, monobasic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.