missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate, Monobasic, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Alle ansehen J.T.Baker Produkte
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
missing translation for 'unitSize'
2.5kg
500g
Spezifikation
Specifications
| Physical Form | Crystals |
| Quantity | 500 g |
| Formula Weight | 136.09 |
| Grade | Meets ACS reagent chemical requirements |
| Packaging | Poly Bottle |
| Specific Gravity | 1L = 2.34kg |
| Chemical Name or Material | Potassium Phosphate, Monobasic |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction