missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate, Monobasic, Ultrapure Bioreagent, J.T. Baker™

Product Code. 15538204 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15538204 2.5 kg Wide Mouth Amber Glass Bottle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15538204 Supplier Avantor J.T.Baker Supplier No. 400805

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Liquid Chromatography and Molecular Biology applications; For Laboratory,Research,or Manufacturing Use

Specifications

CAS 7778-77-0
Physical Form Crystals
Quantity 2.5 kg
Molecular Formula H2KO4P
MDL Number MFCD00011401 MFCD00147253
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES [K+].OP(O)([O-])=O
IUPAC Name potassium;dihydrogen phosphate
Molecular Weight (g/mol) 136.08
PubChem CID 516951
ChEBI CHEBI:63036
Formula Weight 136.09
Grade Ultrapure Bioreagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.34kg
Chemical Name or Material Potassium Phosphate, Monobasic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.