missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate tribasic, 90+%, pure

Product Code. 17231312 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Unit Size:
1kg
25kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17231312 25 kg 25kg
17211312 1 kg 1kg
17221312 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17231312 Supplier Thermo Scientific Acros Supplier No. 467330250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7778-53-2 | K3O4P | 212.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-53-2
Molecular Formula K3O4P
Molecular Weight (g/mol) 212.27
InChI Key LWIHDJKSTIGBAC-UHFFFAOYSA-K
IUPAC Name tripotassium phosphate
SMILES [K+].[K+].[K+].[O-]P([O-])([O-])=O

Specifications

Melting Point 1340.0°C
Quantity 25 kg
Assay Percent Range 90+%
Linear Formula K3PO4
Solubility Information Solubility aq. soln.: 508 g/L (25°C). Other solubilities: insoluble in alcohol
Formula Weight 212.27
Grade Pure
Chemical Name or Material Potassium phosphate tribasic

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.