missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate, tribasic monohydrate, 96%, extra pure

Product Code. 10751331 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Glass bottle
Plastic Bottle
Unit Size:
1kg
25g
25kg
500g
5kg
Product Code. Quantity Packaging unitSize
10751331 25 g Glass bottle 25g
10048521 500 g Glass bottle 500g
10003340 1 kg Glass bottle 1kg
10511984 5 kg Plastic Bottle 5kg
10267123 25 kg Fibre drum 25kg
5 options
This item is not returnable. View return policy

Product Code. 10751331

Brand: Thermo Scientific Acros 263790250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27176-10-9 | K3O4P·H2O | 230.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27176-10-9
Molecular Formula K3O4P·H2O
Molecular Weight (g/mol) 230.28
MDL Number MFCD00150410
InChI Key RMNIZOOYFMNEJJ-UHFFFAOYSA-K
Synonym tri-potassium phosphate monohydrate, potassium phosphate tribasic monohydrate, potassium phosphate hydrate, potassium phosphate monohydrate, unii-4927m96tfv, tert-potassium phosphate monohydrate, potassium phosphate, tribasic, monohydrate, phosphoric acid, tripotassium salt, monohydrate, tripotassium phosphate, monohydrate, acmc-1chx7
PubChem CID 22031701
IUPAC Name tripotassium;phosphate;hydrate
SMILES O.[O-]P(=O)([O-])[O-].[K+].[K+].[K+]

Specifications

Melting Point 1340.0°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 96%
Linear Formula K3PO4·H2O
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol
Formula Weight 230.28
Percent Purity 96%
Grade Extra Pure
Loss on Ignition 13% max. (800°C)
Packaging Glass bottle
Density 2.564
Chemical Name or Material Potassium phosphate, tribasic monohydrate, c.p.
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium phosphate tribasic monohydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.