missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate, Tribasic, n-Hydrate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™
Shop All J.T.Baker ProductsSpecifications
Specifications
| Physical Form | Powder |
| Quantity | 500 g |
| Grade | BAKER ANALYZED™ Reagent |
| Packaging | Poly Bottle |
| Chemical Name or Material | Potassium Phosphate, Tribasic, n-Hydrate |
Safety and Handling
Product Identifier
- Acetonitrile with 0.1% Formic acid
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Acetonitrile, formicacid
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?