missing translation for 'onlineSavingsMsg'
Learn More

Potassium phthalimide, 98+%

Product Code. 11459033
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
2500 g
missing translation for 'unitSize'
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459033 2500 g 2500g
11439033 100 g 100g
11449033 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11459033 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' A11134.0E
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1074-82-4 | C8H4KNO2 | 185.22 g/mol

Potassium phthalimide is used as an intermediate in the synthesis of N-alkylated phthalimides, which is involved in the preparation of primary amines (Gabriel synthesis) by the hydrolysis reaction. It is also used as an intermediate for synthetic indigo, pigments, dyes and pharmaceuticals. Further, it is employed as an organocatalyst for the cyanosilylation of various carbonyl compounds under extremely mild conditions. In addition to this, it serves as a reagent for the transformation of allyl- and alkyl halides into protected primary amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium phthalimide is used as an intermediate in the synthesis of N-alkylated phthalimides, which is involved in the preparation of primary amines (Gabriel synthesis) by the hydrolysis reaction. It is also used as an intermediate for synthetic indigo, pigments, dyes and pharmaceuticals. Further, it is employed as an organocatalyst for the cyanosilylation of various carbonyl compounds under extremely mild conditions. In addition to this, it serves as a reagent for the transformation of allyl- and alkyl halides into protected primary amines.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1074-82-4
Molecular Formula C8H4KNO2
Molecular Weight (g/mol) 185.22
MDL Number MFCD00005887
InChI Key FYRHIOVKTDQVFC-UHFFFAOYSA-M
Synonym potassium phthalimide, potassium 1,3-dioxoisoindolin-2-ide, n-potassiophthalimide, phthalimide potassium salt, n-potassium phthalimide, unii-x6kka27dil, phthalimide, potassium salt, potassium phthalamide, 1h-isoindole-1,3 2h-dione, potassium salt, x6kka27dil
PubChem CID 3356745
IUPAC Name potassium;isoindol-2-ide-1,3-dione
SMILES [K+].O=C1[N-]C(=O)C2=CC=CC=C12

Specifications

Melting Point >300°C
Density 1.63
Quantity 2500 g
Beilstein 3598719
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 185.23
Percent Purity ≥98%
Chemical Name or Material Potassium phthalimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.